Ethidium bromide ≥98% (dry weight)
1 g
Supplier: Thermo Fisher Scientific
Ethidium bromide ≥98% (dry weight)
Catalog # L07482.03
Supplier: Thermo Fisher Scientific
CAS Number: 1239-45-8
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 1239-45-8 Molecular Formula: C21H20BrN3 Formula Weight: 394.32 Odour: Weak Melting Point: ca 261° dec. Beilstein Reference: 3642536 Merck Reference: 14,4731 UN No.: 2811 MDL No.: MFCD00011724
Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
- CAS Number:1239-45-8
- Formula:C₂₁H₂₀BrN₃
- MDL Number:MFCD00011724
- Molecular Weight:394.31 g/mol
- Merck Index:14,4731
- Boiling Pt:72 °C (0.1 mmHg)
- Melting Pt:∼ 261 °C (dec.)
- Density:1.01 g/cm³ (20 °C)
- Flash Pt:<100 °C
- Storage Temperature:Ambient
- EINECS:214-984-6
- UN:2811
- ADR:6.1,I
Documents
Specifications
Specifications
- Pk:1 g



