1,6-Hexanedithiol ≥97%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 1191-43-1 Molecular Formula: HS(CH2)6SH Formula Weight: 150.30 Flash Point: 90°(194°F) Refractive Index: 1.5130 Melting Point: -21° Boiling Point: 242-243° Beilstein Reference: 1732507 UN No.: 2810 MDL No.: MFCD00004910
Synonyms: 1,6-Dimercaptohexane
- CAS Number:1191-43-1
- Formula:C₆H₁₄S₂
- MDL Number:MFCD00004910
- Molecular Weight:150.31 g/mol
- Boiling Pt:242…243 °C
- Melting Pt:–21 °C
- Density:0.990
- Flash Pt:90 °C (194 °F)
- Storage Temperature:Ambient
- EINECS:214-735-1
- UN:3334
- ADR:9,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



