Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,4-Hexanedione 94%

Glass bottle for solids, 100 g
Catalog # L06655.22
Flammable
3,4-Hexanedione 94%
Catalog # L06655.22
CAS Number:  4437-51-8
3,4-Hexanedione 94%
3,4-Hexanedione 94%

Glass bottle for solids, 100 g

CAS Number:  4437-51-8
Price
114,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 94% CAS No.: 4437-51-8 Molecular Formula: C6H10O2 Formula Weight: 114.15 Flash Point: 29°(84°F) Refractive Index: 1.4120 Melting Point: -10° Boiling Point: 129-131° Beilstein Reference: 1700837 UN No.: 1224 MDL No.: MFCD00010237

  • CAS Number:4437-51-8
  • Formula:C₆H₁₀O₂
  • MDL Number:MFCD00010237
  • Molecular Weight:114.14 g/mol
  • Boiling Pt:129…131 °C
  • Melting Pt:–10 °C
  • Density:0.941
  • Flash Pt:29 °C (84 °F)
  • Storage Temperature:Ambient
  • EINECS:224-651-7
  • UN:1224
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications