Order Entry
Export
Orders LinkContactUsLinkComponent
Nessler reagent

Nessler reagent

100 mL
Catalog # LOVI465201
Toxic
Health
Environment
Supplier:   Lovibond
Nessler reagent
Catalog # LOVI465201
Supplier:  Lovibond
CAS Number:  7783-33-7
Nessler reagent

100 mL

CAS Number:  7783-33-7
Price
36,70 €
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution
  • CAS Number:7783-33-7
  • MDL Number:MFCD00049656
  • Boiling Pt:∼100 °C (1013 hPa)
  • Density:1.2 g/cm³ (20 °C)
  • UN:2922
  • ADR:8,II

Documents

Specifications

Specifications

  • Pk:
    100 mL

Specifications