Orders LinkContactUsLinkComponent
Stoppers, with drip tip, MBL®
Stoppers, with drip tip, MBL®
29/32, With pointed bottom
Catalog # 217-0749
Supplier:   SciLabware
Stoppers, with drip tip, MBL®
Catalog # 217-0749
Supplier:  SciLabware
Stoppers, with drip tip, MBL®
Stoppers, with drip tip, MBL®
29/32, With pointed bottom
Price
107,00 €
Log in for availability
1 * 10 items
Product Details & Documents
Ideally suited for use with volumetric flasks etc. Lower closed end of stopper features a 'drip-tip' to ensure adhered liquid is returned to the flask. Flat head allows stopper to be stood on end, thus preventing contamination of the ground surface.
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Hollow-blown to reduce weight
Specifications
  • NS:
    29/32
  • Type:
    With pointed bottom
  • Seq:
    07