Burettes, removable stopcock
Glass burette tube, 50 ml
Catalog # BIBBBRB276P
Supplier: SciLabware
Jump to:
Product Details & Documents
Soda lime glass, class B, removable PTFE stopcock.
- Flared top for safer filling
- Blue enamel graduations
BS 846
Certifications: BS 846
Documents
Specifications
- Description:Glass burette tube, 50 ml
- Seq:04