Orders LinkContactUsLinkComponent
Pouring rings
Pouring rings
Pouring ring, GL 45, Clear
Catalog # 215-3138
Supplier:   SciLabware
Pouring rings
Catalog # 215-3138
Supplier:  SciLabware
Pouring rings
Pouring rings
Pouring ring, GL 45, Clear
Price
10,50 €
Log in for availability
1 * 10 items
Product Details & Documents
Pouring rings, made of Polypropylene, for laboratory/media bottles.
  • Ensures drip-free pouring
  • Very good chemical resistance
  • Autoclavable up to 140 °C
Specifications
  • Thread:
    GL 45
  • Colour:
    Clear
  • Description:
    Pouring ring
  • Seq:
    02