Pouring rings
Pouring ring, GL 32, Clear
BIBB4516/32D
Pouring rings, made of Polypropylene, for laboratory/media bottles.
- Ensures drip-free pouring
- Very good chemical resistance
- Autoclavable up to 140 °C
- Thread:GL 32
- Colour:Clear
- Description:Pouring ring
- Seq:01