Flasks, round bottom, Quickfit® ground joints
250 ml, 14/23 NS, 126 mm
212-0608
PYREX®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
: ISO 1773, ISO 4797.
- Socket size:14/23 NS
- Capacity:250 ml
- Height:126 mm
- Seq:21



