Orders LinkContactUsLinkComponent
Separating funnels, pear shaped, Quickfit®
Separating funnels, pear shaped, Quickfit®
25 ml, 14/23 NS, 14/23 NS
Catalog # 532-0079
Supplier:   SciLabware
Separating funnels, pear shaped, Quickfit®
Catalog # 532-0079
Supplier:  SciLabware
Separating funnels, pear shaped, Quickfit®
Separating funnels, pear shaped, Quickfit®
25 ml, 14/23 NS, 14/23 NS
Price
104,00 €
Log in for availability
1 * 1 items
Product Details & Documents
PYREX®, borosilicate glass.
  • Without graduation
  • Excellent chemical resistance
  • Excellent temperature resistance
Funnel stem with cone to allow easy connection to Quickfit® flask with interchangeable PTFE key stopcock.
Certifications: Complies with ISO 4800 type 2.
Specifications
  • Cone size:
    14/23 NS
  • Socket size:
    14/23 NS
  • Capacity:
    25 ml
  • Seq:
    01