Dropping funnels, cylindrical, Quickfit®
100 ml, 14/23 NS, 14/23 NS
Catalog # 532-0075
Supplier: SciLabware
Jump to:
Product Details & Documents
Funnel stem with ground-glass cone for flasks and assemblies. Stopcock has interchangeable glass key.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Certifications: Complies with ISO 4800
Specifications
- Cone size:14/23 NS
- Socket size:14/23 NS
- Capacity:100 ml
- Seq:01