Orders LinkContactUsLinkComponent
Powder funnels, Quickfit®
34/35 NS, 105 mm
Catalog # 532-0073
Supplier:   SciLabware
Powder funnels, Quickfit®
Catalog # 532-0073
Supplier:  SciLabware
Powder funnels, Quickfit®
Powder funnels, Quickfit®
34/35 NS, 105 mm
Price
45,40 €
Log in for availability
1 * 1 items
Product Details & Documents
PYREX®, borosilicate glass, stem with cone.
  • Very good chemical resistance
  • High temperature resistance
Designed for transfer of powders into flasks and vessels.
Specifications
  • Cone size:
    34/35 NS
  • Funnel O-Ø:
    105 mm
  • Seq:
    05