Orders LinkContactUsLinkComponent
Non return valves, Azlon®
Non return valves, Azlon®
6-8 mm ID
Catalog # AZLONWH902P
Supplier:   SciLabware
Non return valves, Azlon®
Catalog # AZLONWH902P
Supplier:  SciLabware
Non return valves, Azlon®
Non return valves, Azlon®
6-8 mm ID
Price
60,90 €
Log in for availability
1 * 5 items
Product Details & Documents
Designed for use in vacuum lines and fluid handling systems.
  • Useful for preventing back pressure in filter flasks
  • Printed arrow indicates the direction of flow
  • Rigid with stepped tapered arms to give firm grip on tubing
Specifications
  • Tubing size:
    6-8 mm ID