Volumetric flasks, class B, Azlon®
500 ml, 19/26, 0,50 ± ml
Catalog # 214-0247
Supplier: SciLabware
Jump to:
Product Details & Documents
PP, translucent, with PE stopper.
- Class B tolerances with a clear easy to read calibration line
- Feature a mixing chamber at the top of each flask's neck
- Exposure to temperatures above 60 °C may affect accuracy
- Cleaning with mild alkaline detergents recommended up to 60 °C
: ISO 1042
Specifications
- NS:19/26
- Capacity:500 ml
- Tolerance:0,50 ± ml



