Order Entry
Export
Orders LinkContactUsLinkComponent
 

Desiccators, vacuum

With stopcock, 250 DN, 195 mm, 290×296 mm, Vacuum desiccator, with ceramic plate
Catalog # AZLODWA245
Supplier:   SciLabware
Desiccators, vacuum
Catalog # AZLODWA245
Supplier:  SciLabware
Desiccators, vacuum

With stopcock, 250 DN, 195 mm, 290×296 mm, Vacuum desiccator, with ceramic plate

Price
511,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Transparent polycarbonate lid and base.

  • Durable with high impact resistance
  • Safely withstand high vacuum pressures
  • Ceramic plate allows the use of incandescent crucibles

 : Supplied wth plate.

Specifications
  • Nominal size:
    250 DN
  • Type:
    With stopcock
  • Description:
    Vacuum desiccator, with ceramic plate
  • Seq:
    02
  • Usable height:
    195 mm
  • Ø ext.×H:
    290×296 mm