0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Closures
Test/Sample Tube Closures
Stoppers for tubes
Stoppers for tubes
Stoppers for tubes
Grip stopper, 12 mm O.D., Natural
212-0640
:
Greiner Bio-One
Stoppers for tubes
212-0640
:
Greiner Bio-One
Expand image
Stoppers for tubes
Grip stopper, 12 mm O.D., Natural
Add to cart
16,40 €
Unit of Measure
1 * 500 items
-
+
Add to cart
For tubes:
12 mm O.D.
Colour:
Natural
Description:
Grip stopper
Seq:
03