Orders LinkContactUsLinkComponent
Microplates, 96-well, non binding
Microplates, 96-well, non binding
96-well microplate, F-bottom, non binding, chimney well, solid bottom, Clear
Catalog # 738-0170
   Low Manufacturing Impact
Supplier:   Greiner Bio-One
Microplates, 96-well, non binding
Catalog # 738-0170
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Microplates, 96-well, non binding
Microplates, 96-well, non binding
96-well microplate, F-bottom, non binding, chimney well, solid bottom, Clear
Price
277,00 €
Log in for availability
1 * 40 items
Product Details & Documents
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
Specifications
  • Colour:
    Clear
  • Description:
    96-well microplate, F-bottom, non binding, chimney well, solid bottom
  • Seq:
    03
  • Cat. No.:
    738-0170
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Environmentally Preferable:
    Low Manufacturing Impact