Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Modification of configuration of a container added to an existing installation ESP/RCB 1 * 1 items
Catalog # AIRLACCCOOLB60
Supplier:
Air Liquide
Expand image
Modification of configuration of a container added to an existing installation ESP/RCB 1 * 1 items
Catalog # AIRLACCCOOLB60
Supplier:
Air Liquide