Orders LinkContactUsLinkComponent
Counting chambers, Thoma
Counting chambers, Thoma
Counting chamber, bright lines, Thoma
  720-1899
 :   MARIENFELD
Counting chambers, Thoma
  720-1899
Counting chambers, Thoma
Counting chambers, Thoma
Counting chamber, bright lines, Thoma
73,76 €
1 * 1 items
Designed for the determination of the yeast cell number and IVD applications.
Certifications: IVD directive 98/79/EC.
  • Description:
    Counting chamber, bright lines, Thoma
  • Seq:
    3