Orders LinkContactUsLinkComponent
Sheep serum
Sheep serum
Sheep serum, Sterile filtered, Frozen
Catalog # S2350-500
Supplier:   Biowest
Sheep serum
Catalog # S2350-500
Supplier:  Biowest
Sheep serum
Sheep serum
Sheep serum, Sterile filtered, Frozen
Price
72,70 €
Log in for availability
1 * 500 mL
Product Details & Documents
For immunohistochemistry applications, sheep serum is frequently used as a blocking solution. Using sheep serum is useful for blocking the background cross reactivity with immunoglobulins which are already present in the tissue. When used before application of the primary antibody, sheep serum can eliminate Fc receptor binding of the primary and secondary antibody.
Documents
Specifications
  • Serum format:
    Frozen
  • Sterility:
    Sterile filtered
  • Description:
    Sheep serum
  • Seq:
    02