Orders LinkContactUsLinkComponent
Donkey serum
Donkey serum, Sterile filtered, Frozen
Catalog # S2170-500
Supplier:   Biowest
Donkey serum
Catalog # S2170-500
Supplier:  Biowest
Donkey serum
Donkey serum
Donkey serum, Sterile filtered, Frozen
Price
239,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Donkey serum may be used as a blocking agent and controls in immunoassays. For immunohistochemistry and immunocytochemistry, donkey serum was used between 5 to 20% as a blocking agent in various studies.
Specifications
  • Serum format:
    Frozen
  • Sterility:
    Sterile filtered
  • Description:
    Donkey serum
  • Seq:
    25