Donkey serum
Donkey serum, Sterile filtered, Frozen
Supplier: Biowest
Jump to:
Product Details & Documents
Donkey serum may be used as a blocking agent and controls in immunoassays. For immunohistochemistry and immunocytochemistry, donkey serum was used between 5 to 20% as a blocking agent in various studies.
Certificates
Search by Lot NumberSpecifications
- Serum format:Frozen
- Sterility:Sterile filtered
- Description:Donkey serum
- Seq:24
- Cat. No.:S2170-100