96-well plates, polypropylene, without lids
Storage plate, 96-well, expanded volume, natural, V-bottom well, 450 µl, -, 10/bag
: Corning
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or non sterile
: Plates are packaged 25 per pack, 4 packs per case. Nonsterile 450 µL plate is packaged 10 plates per bag and 5 bags per case.
- Packed:10/bag
- Sterile:-
- Description:Storage plate, 96-well, expanded volume, natural, V-bottom well
- Well volume:450 µl
- Cat. No.:732-0484
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Low Manufacturing Impact



