Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
MIDAScan Control Sequence A 1 * 20 µl
MIDAScan Control Sequence A 1 * 20 µl
Print
MIDAScan Control Sequence A 1 * 20 µl
Catalog # TREV4075-020-01
Supplier:
TREVIGEN
MIDAScan Control Sequence A 1 * 20 µl
Catalog # TREV4075-020-01
Supplier:
TREVIGEN
Expand image
Return to Top
MIDAScan Control Sequence A 1 * 20 µl
Add to cart
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 20 µl
-
+
Inquire For Price
Return Policy
Product Details & Documents