Volumetric flasks, class A, blue markings
20** ml, 12/21, 0,040 ± ml
Catalog # 612-5738
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, with glass stopper, conformity certified (DE-M marking).
- Blue enamelled indelible markings resistant to acids and alkalis
- Certificate of conformity with dated batch identification (available via website: www.hirschmannlab.de)
* In addition to DIN
**Wide neck
: DIN EN ISO 1042
Documents
Hirschmann brochure EM-Techcolor -volumetric glassware-
Product Literature
Specifications
- NS:12/21
- Capacity:20** ml
- Seq:06
- Tolerance:0,040 ± ml