Storage Bins, PS
Storage bin, size 5, Grey, 310×485×180 mm
Catalog # HUEN655500
Supplier: hünersdorff GmbH
Jump to:
Product Details & Documents
Storage bins are particularly suitable for a wide range of applications in workshop, store, household and office.
- High load capacity
- Smooth bottom makes it suitable for transport
- Stackable longitudinally and transversely
- Made of PS, coloured
Specifications
- Colour:Grey
- Description:Storage bin, size 5
- Seq:16
- W×D×H:310×485×180 mm



