2,6-Dimethylbenzonitrile
Glass bottle for solids, 1 g
Catalog # MAYBTL00285.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:6575-13-9
- Formula:C₉H₉N
- MDL Number:MFCD00037166
- Molecular Weight:131.18 g/mol
- EINECS:229-503-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



