2,3-Dichlorothiophenol
Glass bottle for solids, 50 g
Catalog # MAYBTL00257.50
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 2,3-Dichlorobenzenethiol, 1,2-Dichloro-3-mercaptobenzene, 2,3-Dichlorobenzene-1-thiol
- CAS Number:17231-95-7
- Formula:C₆H₄Cl₂S
- MDL Number:MFCD00041420
- Molecular Weight:179.07 g/mol
- Boiling Pt:116…118 °C (20 mmHg)
- Melting Pt:46,5 °C
- Melting Pt:46.5 °C
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



