2,5-Dichloronicotinamide
Glass bottle for solids, 1 g
Catalog # MAYBSPB06839.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:75291-86-0
- Formula:C₆H₄Cl₂N₂O
- MDL Number:MFCD00204159
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



