3,5-Dichlorosalicylaldehyde
Glass bottle for solids, 50 g
Catalog # MAYBSB01828.50
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 3,5-Dichloro-2-hydroxybenzaldehyde
- CAS Number:90-60-8
- Formula:C₇H₄Cl₂O₂
- MDL Number:MFCD00003320
- Molecular Weight:191.01 g/mol
- Boiling Pt:723 °C (1013 hPa)
- Melting Pt:92 °C
- EINECS:202-005-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



