Methyl propiolate
Glass bottle for solids, 10 g
MAYBSB01817EA
: Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester
- :922-67-8
- :C₄H₄O₂
- :MFCD00008572
- :84.07 g/mol
- :105 °C (1013 hPa)
- :1.196 g/cm³ (20 °C)
- :213-083-5
- :3272
- :3,II
- Pack type:Glass bottle for solids
- Pk:10 g