Methyl propiolate
Glass bottle for solids, 1 g
Catalog # MAYBSB01817.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester
- CAS Number:922-67-8
- Formula:C₄H₄O₂
- MDL Number:MFCD00008572
- Molecular Weight:84.07 g/mol
- Boiling Pt:105 °C (1013 hPa)
- Density:1.196 g/cm³ (20 °C)
- EINECS:213-083-5
- UN:3272
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



