3,4-Difluorobenzaldehyde
Glass bottle for solids, 10 g
Catalog # MAYBSB01773.10
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:34036-07-2
- Formula:C₇H₄F₂O
- MDL Number:MFCD00010328
- Molecular Weight:142.1 g/mol
- Boiling Pt:53 °C (1013 hPa)
- Density:1,29 g/cm³ (20 °C)
- :Density: 1.29 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:422-180-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



