0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3,4-Difluorobenzaldehyde
3,4-Difluorobenzaldehyde
3,4-Difluorobenzaldehyde
Glass bottle for solids, 1 g
MAYBSB01773.1
:
Thermo Scientific
3,4-Difluorobenzaldehyde
MAYBSB01773.1
:
Thermo Scientific
:
34036-07-2
3,4-Difluorobenzaldehyde
Glass bottle for solids, 1 g
:
34036-07-2
1 * 1 g
-
+
:
34036-07-2
:
C₇H₄F₂O
:
MFCD00010328
:
142.1 g/mol
:
53 °C (1013 hPa)
:
1,29 g/cm³ (20 °C)
:
Density: 1.29 g/cm³ (20 °C)
:
Ambient
:
422-180-3
Pack type:
Glass bottle for solids
Pk:
1 g