5-Bromonicotinic acid
Glass bottle for solids, 50 g
Catalog # MAYBSB01732.50
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 3-Bromo-5-pyridine carboxylic acid, 5-Bromopyridine-3-carboxylic acid, 5-Bromo-3-pyridinecarboxylic acid
- CAS Number:20826-04-4
- Formula:C₆H₄BrNO₂
- MDL Number:MFCD00009783
- Molecular Weight:202.01 g/mol
- Boiling Pt:300 °C (1013 hPa)
- Melting Pt:184…185 °C
- Storage Temperature:Ambient
- EINECS:244-065-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



