4-Quinazolone
Glass bottle for solids, 1 g
Catalog # MAYBSB01696.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 4-Hydroxyquinazoline, Quinazolin-4(3H)-one
- CAS Number:491-36-1
- Formula:C₈H₆N₂O
- MDL Number:MFCD00511302
- Molecular Weight:146.15 g/mol
- EINECS:207-735-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



