Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3,5-Difluorobenzaldehyde
3,5-Difluorobenzaldehyde
3,5-Difluorobenzaldehyde
Glass bottle for solids, 50 g
MAYBSB01678.50
:
Thermo Scientific
3,5-Difluorobenzaldehyde
MAYBSB01678.50
:
Thermo Scientific
:
32085-88-4
3,5-Difluorobenzaldehyde
Glass bottle for solids, 50 g
:
32085-88-4
1 * 50 g
-
+
:
32085-88-4
:
C₇H₄F₂O
:
MFCD00010329
:
142.1 g/mol
:
61…63 °C (43 mmHg)
:
Ambient
:
1989
:
3,III
Pack type:
Glass bottle for solids
Pk:
50 g