3,5-Difluorobenzaldehyde
Glass bottle for solids, 10 g
Catalog # MAYBSB01678.10
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:32085-88-4
- Formula:C₇H₄F₂O
- MDL Number:MFCD00010329
- Molecular Weight:142.1 g/mol
- Boiling Pt:61…63 °C (43 mmHg)
- Storage Temperature:Ambient
- UN:1989
- ADR:3,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



