Sulphamide
Glass bottle for solids, 50 g
Catalog # MAYBSB01649EE
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: Sulphuric diamide
- CAS Number:7803-58-9
- Formula:H₄N₂O₂S
- MDL Number:MFCD00011606
- Molecular Weight:96.11 g/mol
- EINECS:232-262-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



