Order Entry
Export
Orders LinkContactUsLinkComponent
Sulphamide

Sulphamide

Glass bottle for solids, 1 g
Catalog # MAYBSB01649.1
Supplier:   Thermo Scientific
Sulphamide
Catalog # MAYBSB01649.1
Supplier:  Thermo Scientific
CAS Number:  7803-58-9
Sulphamide

Glass bottle for solids, 1 g

CAS Number:  7803-58-9
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Sulphuric diamide
  • CAS Number:7803-58-9
  • Formula:H₄N₂O₂S
  • MDL Number:MFCD00011606
  • Molecular Weight:96.11 g/mol
  • EINECS:232-262-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications