Ethyl propiolate
Glass bottle for solids, 1 g
Catalog # MAYBSB01463.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: Ethyl propynoate, 2-Propynoic acid ethyl ester, Ethyl acetylenecarboxylate, Propiolic acid ethyl ester
- CAS Number:623-47-2
- Formula:C₅H₆O₂
- MDL Number:MFCD00009184
- Molecular Weight:98.1 g/mol
- Boiling Pt:120 °C (1013 hPa)
- Melting Pt:9 °C
- Density:0.9788 g/cm³ (4 °C)
- EINECS:210-795-8
- UN:3272
- ADR:3,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



