4,7-Dichloroquinoline
Glass bottle for solids, 10 g
Catalog # MAYBSB01458.10
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:86-98-6
- Formula:C₉H₅Cl₂N
- MDL Number:MFCD00006774
- Molecular Weight:198.05 g/mol
- Boiling Pt:293 °C (1013 hPa)
- Melting Pt:81…83 °C
- Storage Temperature:Ambient
- EINECS:201-714-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



