3-Methylpyrocatechol
Glass bottle for solids, 10 g
Catalog # MAYBSB01300.10
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 3-Methylcatechol, 3-Methylbenzene-1,2-diol
- CAS Number:488-17-5
- Formula:C₇H₈O₂
- MDL Number:MFCD00016435
- Molecular Weight:124.14 g/mol
- Boiling Pt:241 °C (1013 hPa)
- Melting Pt:58…62 °C
- EINECS:207-672-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



