Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3-Methylpyrocatechol
3-Methylpyrocatechol
3-Methylpyrocatechol
Glass bottle for solids, 1 g
MAYBSB01300.1
:
Thermo Scientific
3-Methylpyrocatechol
MAYBSB01300.1
:
Thermo Scientific
:
488-17-5
3-Methylpyrocatechol
Glass bottle for solids, 1 g
:
488-17-5
1 * 1 g
-
+
:
3-Methylcatechol, 3-Methylbenzene-1,2-diol
:
488-17-5
:
C₇H₈O₂
:
MFCD00016435
:
124.14 g/mol
:
241 °C (1013 hPa)
:
58…62 °C
:
207-672-6
Pack type:
Glass bottle for solids
Pk:
1 g