2-Nitrobenzonitrile
Glass bottle for solids, 10 g
Catalog # MAYBSB01265.10
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: o-Nitrobenzonitrile
- CAS Number:612-24-8
- Formula:C₇H₄N₂O₂
- MDL Number:MFCD00007044
- Molecular Weight:148.12 g/mol
- Boiling Pt:304 °C (1013 hPa)
- Melting Pt:107…109 °C
- Storage Temperature:Ambient
- EINECS:210-301-0
- UN:3439
- ADR:6.1,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



