Order Entry
Export
Orders LinkContactUsLinkComponent
2-Nitrobenzaldehyde

2-Nitrobenzaldehyde

Glass bottle for solids, 50 g
Catalog # MAYBSB01264EE
Supplier:   Thermo Scientific
2-Nitrobenzaldehyde
Catalog # MAYBSB01264EE
Supplier:  Thermo Scientific
CAS Number:  552-89-6
2-Nitrobenzaldehyde

Glass bottle for solids, 50 g

CAS Number:  552-89-6
Price
Inquire For Price
Log in for availability
1 * 50 g
Product Details & Documents
  • CAS Number:552-89-6
  • Formula:C₇H₅NO₃
  • MDL Number:MFCD00007132
  • Molecular Weight:151.12 g/mol
  • Merck Index:12,06684
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:41…43 °C
  • Density:1.35 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:209-025-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications