2-Nitrobenzaldehyde
Glass bottle for solids, 1 g
Catalog # MAYBSB01264.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:552-89-6
- Formula:C₇H₅NO₃
- MDL Number:MFCD00007132
- Molecular Weight:151.12 g/mol
- Merck Index:12,06684
- Boiling Pt:285 °C (1013 hPa)
- Melting Pt:41…43 °C
- Density:1.35 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:209-025-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



