Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2,6-Dibromopyridine
2,6-Dibromopyridine
2,6-Dibromopyridine
Glass bottle for solids, 10 g
MAYBSB01233.10
:
Thermo Scientific
2,6-Dibromopyridine
MAYBSB01233.10
:
Thermo Scientific
:
626-05-1
2,6-Dibromopyridine
Glass bottle for solids, 10 g
:
626-05-1
1 * 10 g
-
+
:
626-05-1
:
C₅H₃Br₂N
:
MFCD00006223
:
236.89 g/mol
:
255 °C (1013 hPa)
:
116…119 °C
:
Ambient
:
210-926-9
:
2811
:
6.1,II
Pack type:
Glass bottle for solids
Pk:
10 g