2-Bromotoluene
Glass bottle for solids, 10 g
Catalog # MAYBSB01220.10
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:95-46-5
- Formula:C₇H₇Br
- MDL Number:MFCD00000068
- Molecular Weight:171.04 g/mol
- Merck Index:13,01424
- Boiling Pt:181.5 °C (1013 hPa)
- Melting Pt:–27 °C
- Density:1.4192 g/cm³ (20 °C)
- Flash Pt:79 °C
- Storage Temperature:Refrigerator
- EINECS:202-421-7
- UN:3082
- ADR:9,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



