6-Chloronicotinic acid
Glass bottle for solids, 50 g
Catalog # MAYBSB01195.50
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 6-Chloropyridine-3-carboxylic acid, 6-Chloro-3-pyridinecarboxylic acid
- CAS Number:5326-23-8
- Formula:C₆H₄ClNO₂
- MDL Number:MFCD00006241
- Molecular Weight:157.56 g/mol
- Boiling Pt:284 °C (1013 hPa)
- Melting Pt:188…189 °C
- Storage Temperature:Ambient
- EINECS:226-201-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



